siRNA Details
| Gene Symbol: PYCR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271681.1 | NP_001258610.1 | 6 | 859 | ||
| NM_013328.3 | NP_037460.2 | 7 | 1081 | |||
| GenBank mRNA | AF087859.1 | - | 7 | 999 | ||
| AF151351.1 | - | 7 | 878 | |||
| AK001508.1 | - | 5 | 2460 | |||
| AK291913.1 | - | 7 | 916 | |||
| AL834217.1 | - | 3 | 814 | |||
| BC014868.1 | - | 7 | 972 | 3919161 | ||
| BC020553.1 | - | 7 | 1006 | 2901360 | ||
| BU538765.1 | - | 6 | 734 | |||
| KU178615.1 | - | 7 | 851 | |||
| KU178616.1 | - | 7 | 675 | |||

PYCR2
Genomic Map