siRNA Details
| Gene Symbol: GMPPB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013334.3 | NP_037466.2 | 5, 6 | 813 | ||
| NM_021971.2 | NP_068806.1 | 5, 6 | 813 | |||
| GenBank mRNA | AF135421.1 | - | 5, 6 | 705 | ||
| AK024319.1 | - | 5, 6 | 790 | |||
| AK291700.1 | - | 5, 6 | 788 | |||
| BC001141.1 | - | 5, 6 | 752 | 3503869 | ||
| BC008033.1 | - | 5, 6 | 752 | 3355099 | ||

GMPPB
Genomic Map