siRNA Details
| Gene Symbol: GMPPA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013335.3 | NP_037467.2 | 5, 6 | 525 | ||
| NM_205847.2 | NP_995319.1 | 5, 6 | 806 | |||
| XM_005246483.3 | XP_005246540.1 | 5, 6 | 525 | |||
| XM_005246486.3 | XP_005246543.1 | 5, 6 | 494 | |||
| XM_011511032.2 | XP_011509334.1 | 5, 6 | 494 | |||
| XM_017003906.1 | XP_016859395.1 | 5, 6 | 525 | |||
| XM_017003907.1 | XP_016859396.1 | 5, 6 | 494 | |||
| XM_017003908.1 | XP_016859397.1 | 5, 6 | 800 | |||
| XM_017003909.1 | XP_016859398.1 | 5, 6 | 525 | |||
| XM_017003910.1 | XP_016859399.1 | 5, 6 | 494 | |||
| XM_017003911.1 | XP_016859400.1 | 5, 6 | 800 | |||
| XM_017003912.1 | XP_016859401.1 | 5, 6 | 525 | |||
| XM_017003913.1 | XP_016859402.1 | 5, 6 | 494 | |||
| XM_017003914.1 | XP_016859403.1 | 5, 6 | 454 | |||
| XM_017003915.1 | XP_016859404.1 | 5, 6 | 684 | |||
| XM_017003916.1 | XP_016859405.1 | 5, 6 | 454 | |||
| XM_017003917.1 | XP_016859406.1 | 5, 6 | 454 | |||
| GenBank mRNA | AB209592.1 | - | 2, 3 | 2159 | ||
| AF135422.2 | - | 4, 5 | 504 | |||
| AK000999.1 | - | 5, 6 | 499 | |||
| AK022578.1 | - | 5, 6 | 800 | |||
| AK222951.1 | - | 5, 6 | 489 | |||
| AK290671.1 | - | 5, 6 | 802 | |||
| AK296193.1 | - | 4, 5 | 752 | |||
| BC007456.2 | - | 5, 6 | 794 | 2989461 | ||
| BP366441.1 | - | 5, 6 | 464 | |||

GMPPA
Genomic Map