siRNA Details
| Gene Symbol: OSGIN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182981.2 | NP_892026.1 | 6 | 1417 | ||
| GenBank mRNA | AF191740.1 | - | 4 | 1054 | ||
| AK290383.1 | - | 6 | 1399 | |||
| AY037158.1 | - | 6 | 1382 | |||
| AY258066.1 | - | 7 | 1868 | |||
| AY258067.1 | - | 6 | 1708 | |||
| AY258068.1 | - | 6 | 1398 | |||
| BC020434.1 | - | 6 | 1383 | |||
| BC032476.1 | - | 6 | 1382 | 5215262 | ||
| BC093687.1 | - | 6 | 1300 | 7939532 | ||
| BC113417.1 | - | 6 | 1300 | 8322469 | ||

OSGIN1
Genomic Map