siRNA Details
| Gene Symbol: G0S2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015714.3 | NP_056529.1 | 2 | 329 | ||
| GenBank mRNA | BC009694.1 | - | 2 | 234 | 3891372 | |
| BM788126.1 | - | 2 | 330 | |||
| BT007101.1 | - | 1 | 72 | |||
| CR542179.1 | - | 1 | 72 | |||
| CR542193.1 | - | 1 | 72 | |||

G0S2
Genomic Map