siRNA Details
| Gene Symbol: ZNF593 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015871.4 | NP_056955.2 | 1, 2 | 290 | ||
| XM_017001398.1 | XP_016856887.1 | 1, 2 | 659 | |||
| GenBank mRNA | AI083892.1 | - | 2, 3 | 360 | ||
| AK311926.1 | - | 1, 2 | 290 | |||
| BC002580.2 | - | 1, 2 | 268 | 3143091 | ||
| BC019267.1 | - | 1, 2 | 263 | 2823658 | ||
| D45213.1 | - | 1, 2 | 199 | |||
| DA124660.1 | - | 1, 2 | 290 | |||

ZNF593
Genomic Map