siRNA Details
| Gene Symbol: MRPS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016034.4 | NP_057118.1 | 4 | 743 | ||
| NR_051967.1 | 5 | 886 | ||||
| NR_051968.1 | 5 | 941 | ||||
| NR_051969.1 | 4 | 946 | ||||
| NR_051970.1 | 4 | 888 | ||||
| XM_006717136.3 | XP_006717199.1 | 5 | 867 | |||
| GenBank mRNA | AB055388.1 | - | 4 | 667 | ||
| AF151849.1 | - | 4 | 667 | |||
| BC004905.2 | - | 1 | 238 | |||
| BC008017.1 | - | 4 | 679 | 3534388 | ||
| BC013108.2 | - | 4 | 687 | 3924671 | ||
| BX362434.2 | - | 4 | 737 | |||

MRPS2
Genomic Map