siRNA Details
Gene Symbol: TCEAL9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006612.1 | NP_001006613.1 | 3 | 586 | ||
NM_001006613.1 | NP_001006614.1 | 2 | 510 | |||
NM_001006614.1 | NP_001006615.1 | 2 | 507 | |||
NM_016303.2 | NP_057387.1 | 3 | 589 | |||
GenBank mRNA | AF125535.1 | - | 3 | 547 | ||
AK312190.1 | - | 3 | 535 | |||
AL833152.1 | - | 1 | 1453 | |||
BC023544.2 | - | 3 | 540 | 4122244 | ||
BQ003038.1 | - | 1 | 510 |
