siRNA Details
| Gene Symbol: MRPL27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016504.2 | NP_057588.1 | 4 | 344 | ||
| GenBank mRNA | AB049647.1 | - | 4 | 319 | ||
| AF151084.1 | - | 4 | 328 | |||
| AK023576.1 | - | 3 | 2058 | |||
| AK315758.1 | - | 4 | 345 | |||
| BC001066.2 | - | 4 | 315 | 2822502 | ||
| BC021986.1 | - | 4 | 329 | 4095663 | ||
| BG829493.1 | - | 4 | 363 | |||
| BM840463.1 | - | 4 | 345 | |||

MRPL27
Genomic Map