siRNA Details
| Gene Symbol: MAPKAPK5-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015404.1 | 2 | 1953 | |||
| GenBank mRNA | AF229831.1 | - | 2 | 458 | ||
| AK096935.1 | - | 2 | 1953 | |||
| AK124158.1 | - | 1 | 1823 | |||
| BC023547.2 | - | 2 | 1604 | |||
| BC136540.1 | - | 2 | 1872 | |||
| JQ794500.1 | - | 2 | 2056 | |||

MAPKAPK5-AS1
Genomic Map