siRNA Details
| Gene Symbol: FKBP11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001143781.1 | NP_001137253.1 | 2 | 153 | ||
| NM_001143782.1 | NP_001137254.1 | 3 | 359 | |||
| NM_016594.2 | NP_057678.1 | 3 | 359 | |||
| GenBank mRNA | AF238079.1 | - | 3 | 312 | ||
| AK094876.1 | - | 1 | 28 | |||
| AK301455.1 | - | 3 | 332 | |||
| AY358998.1 | - | 3 | 317 | |||
| BC027973.1 | - | 3 | 311 | 5229643 | ||
| BC048292.1 | - | 3 | 322 | 5207367 | ||
| BE857354.1 | - | 4 | 378 | |||
| DA378205.1 | - | 3 | 359 | |||

FKBP11
Genomic Map