siRNA Details
| Gene Symbol: TRMO | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016481.4 | NP_057565.3 | 5 | 1304 | ||
| XM_005252048.4 | XP_005252105.1 | 6 | 1364 | |||
| XM_005252049.4 | XP_005252106.1 | 4 | 1086 | |||
| XM_005252050.4 | XP_005252107.1 | 5 | 1190 | |||
| XM_011518777.1 | XP_011517079.1 | 6 | 1343 | |||
| GenBank mRNA | AK000213.1 | - | 5 | 1304 | ||
| AK023069.1 | - | 2 | 2673 | |||
| AY189280.1 | - | 7 | 3357 | |||
| BC002863.2 | - | 5 | 1284 | 3944148 | ||
| X86033.1 | - | 2 | 576 | |||

TRMO
Genomic Map