siRNA Details
| Gene Symbol: SELT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016275.3 | NP_057359.2 | 6 | 1839 | ||
| GenBank mRNA | AK075006.1 | - | 6 | 1812 | ||
| BC026350.1 | - | 6 | 1819 | 4796883 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SELT| Gene Symbol: SELT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016275.3 | NP_057359.2 | 6 | 1839 | ||
| GenBank mRNA | AK075006.1 | - | 6 | 1812 | ||
| BC026350.1 | - | 6 | 1819 | 4796883 | ||