siRNA Details
Gene Symbol: BCL11A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018014.3 | NP_060484.2 | 2 | 554 | ||
NM_022893.3 | NP_075044.2 | 2 | 554 | |||
NM_138559.1 | NP_612569.1 | 2 | 554 | |||
XM_011532909.1 | XP_011531211.1 | 2 | 554 | |||
XM_011532910.1 | XP_011531212.1 | 2 | 554 | |||
XM_011532912.1 | XP_011531214.1 | 2 | 554 | |||
XM_017004333.1 | XP_016859822.1 | 2 | 322 | |||
XM_017004334.1 | XP_016859823.1 | 2 | 554 | |||
XM_017004335.1 | XP_016859824.1 | 2 | 364 | |||
XM_017004336.1 | XP_016859825.1 | 1 | 25 | |||
GenBank mRNA | AB058712.1 | - | 2 | 454 | ||
AF080216.1 | - | 3 | 667 | |||
AK055251.1 | - | 2 | 554 | |||
AK297507.1 | - | 2 | 466 | |||
AK300012.1 | - | 2 | 563 | |||
AY228763.1 | - | 2 | 487 | |||
AY692278.1 | - | 2 | 326 | |||
BC021098.1 | - | 2 | 435 | 5087967 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
