siRNA Details
| Gene Symbol: CHIC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039840.2 | NP_001034929.2 | 6 | 2609 | ||
| NM_001300884.1 | NP_001287813.1 | 5 | 2549 | |||
| XM_017029583.1 | XP_016885072.1 | 7 | 2290 | |||
| GenBank mRNA | AL832095.1 | - | 6 | 2249 | ||
| BC136691.1 | - | 6 | 2514 | 9020681 | ||
| BC136692.1 | - | 6 | 2514 | 9020682 | ||
| CR936642.1 | - | 5 | 2500 | |||

CHIC1
Genomic Map