siRNA Details
| Gene Symbol: HCG4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002139.2 | 1 | 1154 | |||
| GenBank mRNA | AF036977.1 | - | 1 | 1152 | ||
| BC011879.1 | - | 1 | 937 | |||
| Gene Symbol: LOC554223 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_132323.1 | 11 | ||||
| GenBank mRNA | BC051362.1 | - | 11 | 4589959 | ||

HCG4
Genomic Map