siRNA Details
Gene Symbol: HCG4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002139.2 | 1 | 1282 | |||
GenBank mRNA | AF036977.1 | - | 1 | 1280 | ||
BC011879.1 | - | 1 | 1065 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: HCG4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002139.2 | 1 | 1282 | |||
GenBank mRNA | AF036977.1 | - | 1 | 1280 | ||
BC011879.1 | - | 1 | 1065 |