siRNA Details
| Gene Symbol: PRR13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005354.2 | NP_001005354.1 | 3 | 325 | ||
| NM_018457.3 | NP_060927.1 | 3 | 475 | |||
| GenBank mRNA | AF217517.1 | - | 1 | 388 | ||
| AK074398.1 | - | 1 | 371 | |||
| BC001464.2 | - | 3 | 461 | |||
| BC004512.2 | - | 1 | 68 | |||
| BC014257.2 | - | 1 | 375 | 4765821 | ||
| BC016064.1 | - | 1 | 372 | 4852862 | ||
| BC021019.1 | - | 1 | 343 | |||
| BC066943.1 | - | 1 | 374 | 4817871 | ||
| BC071979.1 | - | 1 | 318 | |||
| BX640962.1 | - | 2 | 213 | |||
| CN392914.1 | - | 3 | 489 | |||
| CR533459.1 | - | 2 | 311 | |||

PRR13
Genomic Map