siRNA Details
| Gene Symbol: FAM134B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034850.2 | NP_001030022.1 | 9 | 1178 | ||
| NM_019000.4 | NP_061873.2 | 7 | 1054 | |||
| XM_011514053.2 | XP_011512355.1 | 10 | 1237 | |||
| XM_011514054.2 | XP_011512356.1 | 8 | 1188 | |||
| XM_011514055.2 | XP_011512357.1 | 7 | 978 | |||
| GenBank mRNA | AK000159.1 | - | 1 | 97 | ||
| AK024920.1 | - | 7 | 884 | |||
| AK025808.1 | - | 9 | 1105 | |||
| AK025832.1 | - | 1 | 100 | |||
| AK225371.1 | - | 1 | 100 | |||
| AL832438.1 | - | 9 | 854 | |||
| BC030517.1 | - | 3 | 250 | |||
| BC053326.1 | - | 9 | 1102 | 5014868 | ||
| BC073132.1 | - | 7 | 887 | 5760145 | ||
| CR994722.1 | - | 3 | 293 | |||

FAM134B
Genomic Map