siRNA Details
| Gene Symbol: USP53 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019050.2 | NP_061923.2 | 14 | 2410 | ||
| XM_005263073.3 | XP_005263130.1 | 13 | 2284 | |||
| XM_005263075.2 | XP_005263132.1 | 13 | 2299 | |||
| XM_005263076.2 | XP_005263133.1 | 13 | 2296 | |||
| XM_005263077.3 | XP_005263134.1 | 14 | 2449 | |||
| XM_006714240.1 | XP_006714303.1 | 12 | 1485 | |||
| XM_011532037.2 | XP_011530339.1 | 14 | 2462 | |||
| XM_011532038.2 | XP_011530340.1 | 13 | 1739 | |||
| XM_017008312.1 | XP_016863801.1 | 15 | 2576 | |||
| XM_017008313.1 | XP_016863802.1 | 14 | 2261 | |||
| XM_017008314.1 | XP_016863803.1 | 14 | 2260 | |||
| XM_017008315.1 | XP_016863804.1 | 14 | 2330 | |||
| XM_017008316.1 | XP_016863805.1 | 12 | 2148 | |||
| XM_017008317.1 | XP_016863806.1 | 13 | 2273 | |||
| XM_017008318.1 | XP_016863807.1 | 14 | 2395 | |||
| GenBank mRNA | AB037771.1 | - | 7 | 859 | ||
| AJ583824.1 | - | 11 | 1341 | |||
| AK025301.1 | - | 3 | 311 | |||
| BX648339.1 | - | 12 | 1498 | |||
| CR749490.1 | - | 15 | 2526 | |||

USP53
Genomic Map