siRNA Details
| Gene Symbol: MAP10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019090.2 | NP_061963.2 | 1 | 2873 | ||
| GenBank mRNA | AB037804.1 | - | 1 | 2322 | ||
| AK290216.1 | - | 1 | 2399 | |||
| BC036663.2 | - | 1 | 2351 | 5266476 | ||
| BC109122.2 | - | 1 | 2739 | 40036000 | ||

MAP10
Genomic Map