siRNA Details
| Gene Symbol: ZCCHC10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001300816.2 | NP_001287745.1 | 5 | 613 | ||
| NM_001300817.2 | NP_001287746.1 | 5 | 595 | |||
| NM_001300818.2 | NP_001287747.1 | 4 | 571 | |||
| NM_001300819.2 | NP_001287748.1 | 3 | 505 | |||
| NM_001300822.2 | NP_001287751.1 | 5 | 641 | |||
| NM_001308127.1 | NP_001295056.1 | 5 | 643 | |||
| NM_001308128.1 | NP_001295057.1 | 3 | 385 | |||
| NM_001308129.1 | NP_001295058.1 | 2 | 343 | |||
| NM_001308130.1 | NP_001295059.1 | 4 | 575 | |||
| NM_017665.3 | NP_060135.1 | 4 | 547 | |||
| NR_125337.2 | 4 | 551 | ||||
| NR_131766.1 | 5 | 696 | ||||
| NR_131767.1 | 4 | 579 | ||||
| XM_011543502.1 | XP_011541804.1 | 4 | 600 | |||
| XM_011543503.2 | XP_011541805.1 | 5 | 688 | |||
| GenBank mRNA | AF086446.1 | - | 3 | 429 | ||
| AK000101.1 | - | 4 | 491 | |||
| AK122970.1 | - | 5 | 540 | |||
| AK300543.1 | - | 3 | 473 | |||
| BC005211.1 | - | 4 | 504 | 3961163 | ||
| BC022443.1 | - | 5 | 554 | 4285562 | ||
| BF983215.1 | - | 4 | 509 | |||
| BI766652.1 | - | 2 | 275 | |||
| BI826979.1 | - | 4 | 547 | |||
| BM470321.1 | - | 4 | 498 | |||
| BU165290.1 | - | 4 | 502 | |||
| CB112418.1 | - | 5 | 582 | |||
| DA345717.1 | - | 3 | 353 | |||
| DA528172.1 | - | 4 | 519 | |||

ZCCHC10
Genomic Map