siRNA Details
| Gene Symbol: TMEM161A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001256766.1 | NP_001243695.1 | 7, 8 | 677 | ||
| NM_017814.2 | NP_060284.1 | 9, 10 | 986 | |||
| XM_011528117.2 | XP_011526419.1 | 4, 5 | 747 | |||
| XM_017026916.1 | XP_016882405.1 | 9, 10 | 882 | |||
| GenBank mRNA | AK000429.1 | - | 9, 10 | 943 | ||
| AK096964.1 | - | 7, 8 | 642 | |||
| AK225424.1 | - | 9, 10 | 900 | |||
| AY358088.1 | - | 9, 10 | 970 | |||
| BC005210.1 | - | 9, 10 | 946 | 4041226 | ||
| BC016478.2 | - | 8, 9 | 910 | |||

TMEM161A
Genomic Map