siRNA Details
| Gene Symbol: TMEM160 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017854.1 | NP_060324.1 | 1, 2 | 220 | ||
| GenBank mRNA | AK000519.1 | - | 1, 2 | 220 | ||
| BC002907.2 | - | 1, 2 | 247 | 3941377 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TMEM160| Gene Symbol: TMEM160 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017854.1 | NP_060324.1 | 1, 2 | 220 | ||
| GenBank mRNA | AK000519.1 | - | 1, 2 | 220 | ||
| BC002907.2 | - | 1, 2 | 247 | 3941377 | ||