siRNA Details
| Gene Symbol: ODAM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017855.3 | NP_060325.3 | 7, 8 | 586 | ||
| XM_005265698.3 | XP_005265755.1 | 8, 9 | 650 | |||
| XM_017008331.1 | XP_016863820.1 | 6, 7 | 542 | |||
| GenBank mRNA | AK000520.1 | - | 4, 5 | 237 | ||
| AK225547.1 | - | 4, 5 | 217 | |||
| BC017796.1 | - | 3, 4 | 193 | 4689080 | ||
| BG194365.1 | - | 7, 8 | 589 | |||
| EF113908.1 | - | 7, 8 | 588 | |||

ODAM
Genomic Map