siRNA Details
| Gene Symbol: SOBP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018013.3 | NP_060483.3 | 3 | 879 | ||
| XM_005267041.4 | XP_005267098.1 | 3 | 887 | |||
| XM_005267042.4 | XP_005267099.1 | 3 | 888 | |||
| XM_011535920.2 | XP_011534222.1 | 3 | 887 | |||
| XM_011535921.2 | XP_011534223.1 | 3 | 411 | |||
| XM_011535923.2 | XP_011534225.1 | 1 | 26 | |||
| XM_017010991.1 | XP_016866480.1 | 2 | 1402 | |||
| XM_017010994.1 | XP_016866483.1 | 1 | 29 | |||
| GenBank mRNA | AK090879.1 | - | 3 | 483 | ||
| BC091526.1 | - | 3 | 388 | |||
| DQ507800.1 | - | 3 | 375 | |||

SOBP
Genomic Map