siRNA Details
| Gene Symbol: HEATR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018072.5 | NP_060542.4 | 38 | 5619 | ||
| GenBank mRNA | AK001221.1 | - | 2 | 139 | ||
| AK098212.1 | - | 13 | 1890 | |||
| AK125663.1 | - | 20 | 2836 | |||
| AK304129.1 | - | 18 | 2696 | |||
| AK316413.1 | - | 18 | 2590 | |||
| BC011983.1 | - | 2 | 233 | |||
| BC035302.1 | - | 9 | 1194 | |||
| BC065205.1 | - | 16 | 2355 | |||
| BC130000.1 | - | 23 | 3543 | |||
| BC150614.1 | - | 37 | 5489 | 9056984 | ||

HEATR1
Genomic Map