siRNA Details
| Gene Symbol: THAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018105.2 | NP_060575.1 | 3 | 756 | ||
| NM_199003.1 | NP_945354.1 | 2 | 560 | |||
| GenBank mRNA | AK001339.1 | - | 3 | 750 | ||
| AK223231.1 | - | 3 | 735 | |||
| AL832077.1 | - | 2 | 534 | |||
| BC021721.2 | - | 3 | 730 | 4795229 | ||
| CD639319.1 | - | 2 | 542 | |||

THAP1
Genomic Map