siRNA Details
| Gene Symbol: THAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018105.2 | NP_060575.1 | 3 | 865 | ||
| NM_199003.1 | NP_945354.1 | 2 | 669 | |||
| GenBank mRNA | AK001339.1 | - | 3 | 859 | ||
| AL832077.1 | - | 2 | 643 | |||
| BC021721.2 | - | 3 | 839 | 4795229 | ||

THAP1
Genomic Map