siRNA Details
Gene Symbol: TMEM100 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099640.1 | NP_001093110.1 | 4 | 892 | ||
NM_018286.2 | NP_060756.2 | 2 | 394 | |||
XM_017024814.1 | XP_016880303.1 | 5 | 2249 | |||
XM_017024815.1 | XP_016880304.1 | 3 | 358 | |||
XM_017024816.1 | XP_016880305.1 | 2 | 410 | |||
GenBank mRNA | AK001832.1 | - | 2 | 311 | ||
BC010128.2 | - | 2 | 244 | 3952255 | ||
DA896013.1 | - | 2 | 394 |
