siRNA Details
| Gene Symbol: SLC35C1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145265.1 | NP_001138737.1 | 3 | 1394 | ||
| NM_001145266.1 | NP_001138738.1 | 3 | 1297 | |||
| NM_018389.4 | NP_060859.4 | 2 | 1668 | |||
| XM_011520202.2 | XP_011518504.1 | 2 | 555 | |||
| GenBank mRNA | AF323970.1 | - | 2 | 956 | ||
| AF326199.1 | - | 2 | 1565 | |||
| AK000527.1 | - | 1 | 213 | |||
| AK002182.1 | - | 3 | 1070 | |||
| AK027394.1 | - | 2 | 1645 | |||
| AK074929.1 | - | 2 | 1669 | |||
| AK091245.1 | - | 2 | 2352 | |||
| AK225428.1 | - | 3 | 1092 | |||
| AK315473.1 | - | 2 | 1370 | |||
| BC001427.2 | - | 2 | 973 | 3139599 | ||

SLC35C1
Genomic Map