siRNA Details
| Gene Symbol: RBM38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001291780.1 | NP_001278709.1 | 1 | 270 | ||
| NM_017495.5 | NP_059965.2 | 1 | 270 | |||
| NM_183425.2 | NP_906270.1 | 1 | 270 | |||
| XM_005260446.3 | XP_005260503.1 | 1 | 208 | |||
| XM_011528885.2 | XP_011527187.1 | 1 | 202 | |||
| GenBank mRNA | AF432218.1 | - | 1 | 266 | ||
| BC018711.1 | - | 1 | 22 | 4852523 | ||
| BM549757.1 | - | 265 | ||||
| DN998561.1 | - | 1 | 255 | |||

RBM38
Genomic Map