siRNA Details
| Gene Symbol: ZNF823 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080493.3 | NP_001073962.1 | 4 | 1217 | ||
| NM_001297610.1 | NP_001284539.1 | 3 | 1156 | |||
| NM_017507.1 | NP_059977.1 | 3 | 1090 | |||
| XM_017026941.1 | XP_016882430.1 | 4 | 1379 | |||
| XM_017026942.1 | XP_016882431.1 | 2 | 1017 | |||
| GenBank mRNA | AK300579.1 | - | 4 | 1184 | ||
| AK303213.1 | - | 3 | 1121 | |||
| AK308189.1 | - | 3 | 1026 | |||
| BC052597.1 | - | 4 | 1129 | 4304302 | ||
| BC063560.1 | - | 1 | 390 | |||
| X51760.1 | - | 4 | 1131 | |||

ZNF823
Genomic Map