siRNA Details
| Gene Symbol: RNF126 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_194460.2 | NP_919442.1 | 9 | 1384 | ||
| XM_005259594.1 | XP_005259651.1 | 9 | 1383 | |||
| GenBank mRNA | AL360265.1 | - | 4 | 1830 | ||
| BC001442.2 | - | 10 | 1156 | 3138961 | ||
| BG773910.1 | - | 1 | 149 | |||
| CA440294.1 | - | 1 | 277 | |||

RNF126
Genomic Map