siRNA Details
| Gene Symbol: NECAP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145277.1 | NP_001138749.1 | 2 | 232 | ||
| NM_001145278.1 | NP_001138750.1 | 2 | 154 | |||
| NM_018090.4 | NP_060560.1 | 2 | 232 | |||
| XM_011541767.2 | XP_011540069.1 | 3 | 304 | |||
| XM_017001721.1 | XP_016857210.1 | 2 | 180 | |||
| GenBank mRNA | AK001282.1 | - | 2 | 164 | ||
| AK023545.1 | - | 2 | 168 | |||
| AK024468.1 | - | 2 | 146 | |||
| AK223344.1 | - | 2 | 180 | |||
| AK300764.1 | - | 2 | 178 | |||
| AK302227.1 | - | 2 | 100 | |||
| BC017014.1 | - | 2 | 163 | 3848563 | ||
| BC018914.2 | - | 2 | 166 | 4299890 | ||
| BM751287.1 | - | 2 | 232 | |||

NECAP2
Genomic Map