siRNA Details
Gene Symbol: ERMARD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278532.1 | NP_001265461.1 | 15 | 1457 | ||
NM_018341.2 | NP_060811.1 | 16 | 1626 | |||
XM_011535938.2 | XP_011534240.1 | 15 | 1596 | |||
XM_011535939.2 | XP_011534241.1 | 16 | 2716 | |||
XM_011535940.2 | XP_011534242.1 | 16 | 2525 | |||
XM_017011027.1 | XP_016866516.1 | 16 | 2466 | |||
XM_017011028.1 | XP_016866517.1 | 16 | 1791 | |||
XM_017011029.1 | XP_016866518.1 | 16 | 1551 | |||
XM_017011030.1 | XP_016866519.1 | 15 | 1375 | |||
XM_017011031.1 | XP_016866520.1 | 15 | 1440 | |||
XM_017011032.1 | XP_016866521.1 | 16 | 2711 | |||
XM_017011033.1 | XP_016866522.1 | 16 | 2520 | |||
GenBank mRNA | AK002014.1 | - | 16 | 1623 | ||
AK294090.1 | - | 15 | 1454 |
