siRNA Details
| Gene Symbol: ZMAT5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001003692.1 | NP_001003692.1 | 5 | 425 | ||
| NM_001318129.1 | NP_001305058.1 | 5 | 379 | |||
| NM_019103.2 | NP_061976.1 | 6 | 540 | |||
| GenBank mRNA | AA312132.1 | - | 5 | 425 | ||
| AK292671.1 | - | 5 | 406 | |||
| AL160311.1 | - | 5 | 424 | |||
| BC009717.1 | - | 5 | 390 | 3890730 | ||
| BQ931527.1 | - | 5 | 353 | |||
| BU598307.1 | - | 5 | 427 | |||
| CR456353.1 | - | 4 | 312 | |||
| CR457393.1 | - | 4 | 289 | |||

ZMAT5
Genomic Map