siRNA Details
Gene Symbol: YLPM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019589.2 | NP_062535.2 | 5 | 2464 | ||
XM_005267860.3 | XP_005267917.1 | 5 | 2508 | |||
XM_011536966.2 | XP_011535268.1 | 6 | 1623 | |||
GenBank mRNA | AK090435.1 | - | 3 | 1220 | ||
AL832365.1 | - | 2 | 999 | |||
L40400.1 | - | 2 | 143 |
