siRNA Details
| Gene Symbol: KCNK12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022055.1 | NP_071338.1 | 2 | 873 | ||
| GenBank mRNA | AF287302.1 | - | 2 | 873 | ||
| BC047749.1 | - | 2 | 901 | 6186991 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

KCNK12| Gene Symbol: KCNK12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022055.1 | NP_071338.1 | 2 | 873 | ||
| GenBank mRNA | AF287302.1 | - | 2 | 873 | ||
| BC047749.1 | - | 2 | 901 | 6186991 | ||