siRNA Details
| Gene Symbol: FMN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001305424.1 | NP_001292353.1 | 17 | 5165 | ||
| NM_020066.4 | NP_064450.3 | 16 | 5153 | |||
| XM_017001839.1 | XP_016857328.1 | 17 | 4005 | |||
| XM_017001840.1 | XP_016857329.1 | 16 | 3405 | |||
| XM_017001841.1 | XP_016857330.1 | 16 | 3863 | |||
| XM_017001842.1 | XP_016857331.1 | 15 | 3084 | |||
| XM_017001843.1 | XP_016857332.1 | 14 | 3072 | |||
| GenBank mRNA | AB209153.1 | - | 17 | 3758 | ||
| AF218942.1 | - | 8 | 653 | |||
| AF225426.1 | - | 8 | 675 | |||
| AK297716.1 | - | 11 | 923 | |||
| AK298141.1 | - | 9 | 988 | |||
| BC014364.2 | - | 6 | 492 | |||

FMN2
Genomic Map