siRNA Details
| Gene Symbol: SMCO4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020179.2 | NP_064564.1 | 3 | 346 | ||
| XM_011542907.1 | XP_011541209.1 | 3 | 371 | |||
| XM_011542908.2 | XP_011541210.1 | 4 | 474 | |||
| XM_011542909.1 | XP_011541211.1 | 3 | 356 | |||
| XM_011542910.2 | XP_011541212.1 | 3 | 1109 | |||
| XM_011542911.2 | XP_011541213.1 | 3 | 1035 | |||
| XM_017018019.1 | XP_016873508.1 | 3 | 1091 | |||
| XM_017018020.1 | XP_016873509.1 | 4 | 532 | |||
| GenBank mRNA | AF197137.1 | - | 3 | 318 | ||

SMCO4
Genomic Map