siRNA Details
| Gene Symbol: ATXN7L3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001098833.1 | NP_001092303.1 | 3 | 420 | ||
| NM_020218.1 | NP_064603.1 | 3 | 420 | |||
| XM_011525038.2 | XP_011523340.1 | 3 | 1014 | |||
| XM_011525044.2 | XP_011523346.1 | 3 | 1014 | |||
| XM_011525046.2 | XP_011523348.1 | 3 | 1014 | |||
| XM_017024880.1 | XP_016880369.1 | 4 | 592 | |||
| XM_017024881.1 | XP_016880370.1 | 4 | 891 | |||
| XM_017024882.1 | XP_016880371.1 | 3 | 1014 | |||
| XM_017024883.1 | XP_016880372.1 | 4 | 595 | |||
| XM_017024884.1 | XP_016880373.1 | 3 | 638 | |||
| XM_017024885.1 | XP_016880374.1 | 4 | 595 | |||
| XM_017024886.1 | XP_016880375.1 | 4 | 595 | |||
| GenBank mRNA | AK126117.1 | - | 3 | 638 | ||
| BC037418.1 | - | 3 | 327 | |||
| BC113595.1 | - | 3 | 420 | 8322647 | ||
| BC126113.1 | - | 3 | 420 | 8991829 | ||

ATXN7L3
Genomic Map