siRNA Details
| Gene Symbol: TOMM22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020243.4 | NP_064628.1 | 1, 2 | 155 | ||
| GenBank mRNA | AB040119.1 | - | 1, 2 | 137 | ||
| AB041906.1 | - | 1, 2 | 138 | |||
| AF163256.1 | - | 1, 2 | 138 | |||
| AK024731.1 | - | 1, 2 | 148 | |||
| AK223021.1 | - | 1, 2 | 153 | |||
| AK312076.1 | - | 1, 2 | 129 | |||
| BC009363.2 | - | 1, 2 | 130 | 4302531 | ||
| CR456475.1 | - | 1, 2 | 130 | |||

TOMM22
Genomic Map