siRNA Details
| Gene Symbol: ANKMY2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020319.2 | NP_064715.1 | 7 | 1015 | ||
| XM_011515458.1 | XP_011513760.1 | 8 | 1142 | |||
| GenBank mRNA | AK001389.1 | - | 8 | 984 | ||
| AK001740.1 | - | 7 | 953 | |||
| AK023704.1 | - | 7 | 952 | |||
| AL050390.1 | - | 7 | 956 | |||
| BC035353.1 | - | 7 | 971 | 5185501 | ||

ANKMY2
Genomic Map