siRNA Details
| Gene Symbol: RGL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001035223.3 | NP_001030300.3 | 19 | 2144 | ||
| NM_001161616.2 | NP_001155088.2 | 19 | 2162 | |||
| GenBank mRNA | AK096811.1 | - | 13 | 1749 | ||
| AK126263.1 | - | 1 | 1682 | |||
| BC014426.1 | - | 19 | 2130 | |||
| BC064475.1 | - | 4 | 1161 | |||
| BC101756.1 | - | 19 | 2130 | 8069262 | ||
| BC111958.1 | - | 19 | 2130 | 8327426 | ||
| BC143530.1 | - | 19 | 2148 | 9052039 | ||
| BM992649.1 | - | 1 | 414 | |||

RGL3
Genomic Map