siRNA Details
| Gene Symbol: SMIM8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001042493.2 | NP_001035958.1 | 3 | 205 | ||
| NM_001287445.1 | NP_001274374.1 | 2 | 184 | |||
| NM_020425.5 | NP_065158.3 | 2 | 184 | |||
| GenBank mRNA | AF311339.1 | - | 2 | 226 | ||
| AK311963.1 | - | 3 | 176 | |||
| AL050201.1 | - | 1 | 151 | |||
| BC070260.1 | - | 2 | 166 | 6452586 | ||
| BF212252.1 | - | 163 | ||||
| BG534658.1 | - | 2 | 185 | |||
| DA293899.1 | - | 3 | 176 | |||

SMIM8
Genomic Map