siRNA Details
| Gene Symbol: SMIM8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001042493.2 | NP_001035958.1 | 4 | 241 | ||
| NM_020425.5 | NP_065158.3 | 3 | 220 | |||
| GenBank mRNA | AF311339.1 | - | 3 | 262 | ||
| AK311963.1 | - | 4 | 212 | |||
| AL050201.1 | - | 2 | 187 | |||
| BC070260.1 | - | 3 | 202 | 6452586 | ||
| BG534658.1 | - | 3 | 221 | |||
| DA293899.1 | - | 4 | 212 | |||

SMIM8
Genomic Map