siRNA Details
| Gene Symbol: GJC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020435.3 | NP_065168.2 | 2 | 457 | ||
| GenBank mRNA | AY285161.1 | - | 1 | 135 | ||
| BC035840.1 | - | 2 | 211 | |||
| BC089439.1 | - | 2 | 457 | 30558819 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

GJC2| Gene Symbol: GJC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020435.3 | NP_065168.2 | 2 | 457 | ||
| GenBank mRNA | AY285161.1 | - | 1 | 135 | ||
| BC035840.1 | - | 2 | 211 | |||
| BC089439.1 | - | 2 | 457 | 30558819 | ||