siRNA Details
| Gene Symbol: PPM1H | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020700.1 | NP_065751.1 | 3 | 889 | ||
| XM_011538578.2 | XP_011536880.1 | 3 | 1025 | |||
| XM_011538579.1 | XP_011536881.1 | 3 | 577 | |||
| XM_017019676.1 | XP_016875165.1 | 3 | 1139 | |||
| GenBank mRNA | AB032983.1 | - | 2 | 482 | ||
| AB084258.1 | - | 3 | 1154 | |||
| AK023315.1 | - | 4 | 951 | |||
| BC157843.1 | - | 3 | 899 | 9057062 | ||

PPM1H
Genomic Map