siRNA Details
| Gene Symbol: SHROOM4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020717.3 | NP_065768.2 | 3 | 415 | ||
| NR_027121.1 | 3 | 415 | ||||
| XM_017029682.1 | XP_016885171.1 | 3 | 521 | |||
| XM_017029684.1 | XP_016885173.1 | 2 | 538 | |||
| XM_017029685.1 | XP_016885174.1 | 3 | 538 | |||
| XM_017029687.1 | XP_016885176.1 | 3 | 525 | |||
| GenBank mRNA | AB033028.1 | - | 3 | 415 | ||
| AY044234.1 | - | 2 | 496 | |||
| BC151240.1 | - | 3 | 426 | 8860513 | ||

SHROOM4
Genomic Map